Learn More
Ethyl isobutyrate, 98%
Product Code. 11439823
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439823 100 mL 100mL
11449823 500 mL 500mL
11459823 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11439823 Supplier Thermo Scientific Alfa Aesar Supplier No. A12966.AE
Request Bulk or Custom Format
CAS: 97-62-1 | C6H12O2 | 116.16 g/mol
It is employed to make flavoring compounds and essences, as a flavoring agent and to flavor animal feeds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed to make flavoring compounds and essences, as a flavoring agent and to flavor animal feeds.
Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from ignition sources. Keep away from electrostatic charges.
TRUSTED_SUSTAINABILITY
Identificadores de productos químicos
CAS 97-62-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009165
InChI Key WDAXFOBOLVPGLV-UHFFFAOYSA-N
Synonym ethyl isobutyrate, ethyl isobutanoate, ethylisobutyrate, isobutyric acid ethyl ester, ethyl 2,2-dimethylacetate, propanoic acid, 2-methyl-, ethyl ester, isobutyric acid, ethyl ester, ethyl 2-methylpropionate, propionic acid, 2-methyl-, ethyl ester, ethyl isobutyrate natural
PubChem CID 7342
ChEBI CHEBI:87303
IUPAC Name ethyl 2-methylpropanoate
SMILES CCOC(=O)C(C)C
Especificaciones
Melting Point -88°C
Density 0.869
Boiling Point 108°C to 110°C
Flash Point 13°C (55°F)
Odor Pleasant
Refractive Index 1.387
Quantity 100 mL
UN Number UN2385
Beilstein 773846
Merck Index 14,3814
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol.
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material Ethyl isobutyrate
Mostrar más Mostrar menos
RUO – Research Use Only
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.