missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isonicotinate, 98%

Product Code. 11337935
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337935 100 g 100g
11347935 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337935 Supplier Thermo Scientific Alfa Aesar Supplier No. L03010.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1570-45-2 | C8H9NO2 | 151.165 g/mol

Ethyl isonicotinate is an organic building block, used in the synthesis of various pharmaceutical and biologically active compounds. It is also a pyridine compound that is shown to be able to trap the thrips in the corp.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl isonicotinate is an organic building block, used in the synthesis of various pharmaceutical and biologically active compounds. It is also a pyridine compound that is shown to be able to trap the thrips in the corp.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1570-45-2
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00006428
InChI Key MCRPKBUFXAKDKI-UHFFFAOYSA-N
Synonym ethyl isonicotinate, 4-picolinic acid ethyl ester, ethyl 4-pyridinecarboxylate, isonicotinic acid, ethyl ester, 4-carbethoxypyridine, 4-pyridinecarboxylic acid, ethyl ester, isonicotinic acid ethyl ester, 4-carboethoxypyridine, gamma-pyridinecarboxylic acid ethyl ester, .gamma.-pyridinecarboxylic acid ethyl ester
PubChem CID 15291
IUPAC Name ethyl pyridine-4-carboxylate
SMILES CCOC(=O)C1=CC=NC=C1

Specifications

Density 1.008
Boiling Point 220°C
Flash Point 87°C (188°F)
Odor Characteristic
Refractive Index 1.501
Quantity 100 g
Beilstein 122942
Merck Index 14,5187
Solubility Information Slightly soluble in water.
Formula Weight 151.17
Percent Purity 98%
Chemical Name or Material Ethyl isonicotinate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.