Learn More
Ethyl isonicotinate, 98%
Description
Ethyl isonicotinate is an organic building block, used in the synthesis of various pharmaceutical and biologically active compounds. It is also a pyridine compound that is shown to be able to trap the thrips in the corp.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethyl isonicotinate is an organic building block, used in the synthesis of various pharmaceutical and biologically active compounds. It is also a pyridine compound that is shown to be able to trap the thrips in the corp.
Solubility
Slightly soluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Specifications
| Density | 1.008 |
| Boiling Point | 220°C |
| Flash Point | 87°C (188°F) |
| Odor | Characteristic |
| Refractive Index | 1.501 |
| Quantity | 500 g |
| Beilstein | 122942 |
| Merck Index | 14,5187 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 151.17 |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.