missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Laurate(Ethyl Dodecanoate), TRC

Product Code. 30379008
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30379008 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30379008 Supplier Toronto Research Chemicals Supplier No. TRCE9173855G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lauric Acid Ethyl Ester
CAS 106-33-2
Molecular Formula C14 H28 O2
Quantity 5 g
Synonym Dodecanoic Acid Ethyl Ester, Dodecanoic acid, ethyl ester, Lauric acid, ethyl ester (6CI,8CI), Dodecanoic acid monoethyl ester, Ethyl dodecanoate, Ethyl dodecylate, Ethyl laurate, Ethyl laurinate, Ethyl n-dodecanoate, NSC 83467, NSC 8912
InChI Formula InChI=1S/C14H28O2/c1-3-5-6-7-8-9-10-11-12-13-14(15)16-4-2/h3-13H2,1-2H3
SMILES CCCCCCCCCCCC(=O)OCC
IUPAC Name ethyl dodecanoate
Molecular Weight (g/mol) 228.37
Formula Weight 228.2089
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.