missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Maltol, Mikromol™

Product Code. 17932146
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17932146 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17932146 Supplier Mikromol Supplier No. MM3722.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ethyl Maltol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl maltol
CAS 11-8-4940
Analyte or Component Names Ethyl maltol
Shipping Condition Room Temperature
Molecular Formula C7 H8 O3
Certifications/Compliance ISO 17034
Synonym 4H-Pyran-4-one, 2-ethyl-3-hydroxy-, 2-Ethyl-3-hydroxy-4-pyrone, 2-Ethyl-3-hydroxy-4H-pyran-4-one, 2-Ethylpyromeconic acid, 3-Hydroxy-2-ethyl-1,4-pyrone, 3-Hydroxy-2-ethyl-4-pyrone, 3-Hydroxy-2-ethyl-γ-pyrone, Ethyl maltol, NSC 638851, Veltol plus
InChI Formula InChI=1S/C7H8O3/c1-2-6-7(9)5(8)3-4-10-6/h3-4,9H,2H2,1H3
SMILES CCC1=C(O)C(=O)C=CO1
IUPAC Name 2-ethyl-3-hydroxypyran-4-one
Molecular Weight (g/mol) 140.14
Quantity 250 mg
Formula Weight 140.0473 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.