missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Maltol, TRC

Product Code. 17954455
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17954455 25 g 25g
17962705 10 g 10g
17934295 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17954455 Supplier Toronto Research Chemicals Supplier No. TRCE92200525G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4940-11-8
Molecular Formula C7 H8 O3
Molecular Weight (g/mol) 140.14
Synonym 4H-Pyran-4-one, 2-ethyl-3-hydroxy-, 2-Ethyl-3-hydroxy-4-pyrone, 2-Ethyl-3-hydroxy-4H-pyran-4-one, 2-Ethylpyromeconic acid, 3-Hydroxy-2-ethyl-1,4-pyrone, 3-Hydroxy-2-ethyl-4-pyrone, 3-Hydroxy-2-ethyl-γ-pyrone, Ethyl maltol, NSC 638851, Veltol plus
IUPAC Name 2-ethyl-3-hydroxypyran-4-one
SMILES CCC1=C(O)C(=O)C=CO1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C7H8O3/c1-2-6-7(9)5(8)3-4-10-6/h3-4,9H,2H2,1H3
Formula Weight 140.0473
Chemical Name or Material Ethyl maltol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.