missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Myristate, TRC

Product Code. 30383642
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30383642 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383642 Supplier Toronto Research Chemicals Supplier No. TRCE925180100MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetradecanoic acid ethyl ester
CAS 124-06-1
Molecular Formula C16 H32 O2
Quantity 100 mg
Synonym Tetradecanoic acid, ethyl ester (9CI, ACI), Myristic acid, ethyl ester (6CI, 7CI, 8CI), Ethyl myristate, Ethyl tetradecanoate, Nikkol BM, NSC 8917
InChI Formula InChI=1S/C16H32O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16(17)18-4-2/h3-15H2,1-2H3
SMILES CCCCCCCCCCCCCC(=O)OCC
IUPAC Name ethyl tetradecanoate
Molecular Weight (g/mol) 256.42
Formula Weight 256.2402
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.