missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Octanoate, TRC

Product Code. 30385506
Change view
Click to view available options
Quantity:
25 mL
50 mL
Unit Size:
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30385506 25 mL -
30382657 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30385506 Supplier Toronto Research Chemicals Supplier No. TRCE92531525ML
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-32-1
Molecular Formula C10 H20 O2
Molecular Weight (g/mol) 172.26
Synonym Octanoic acid-ethyl ester, Octanoic acid, ethyl ester (9CI, ACI), Caprylic acid ethyl ester, Et caprylate, Ethyl caprylate, Ethyl n-octanoate, Ethyl octanoate, NSC 8898
IUPAC Name ethyl octanoate
SMILES CCCCCCCC(=O)OCC

Specifications

Quantity 25 mL
InChI Formula InChI=1S/C10H20O2/c1-3-5-6-7-8-9-10(11)12-4-2/h3-9H2,1-2H3
Formula Weight 172.1463
Chemical Name or Material Caprylic acid ethyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.