missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Palmitate, TRC

Product Code. 17929994
Change view
Click to view available options
Quantity:
500 mg
1 g
100 g
Unit Size:
100g
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17929994 500 mg 500mg
17978974 100 g 100g
17969984 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17929994 Supplier Toronto Research Chemicals Supplier No. TRCE925480500MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-97-7
Molecular Formula C18 H36 O2
Molecular Weight (g/mol) 284.48
Synonym Hexadecanoic acid, ethyl ester (9CI, ACI), Palmitic acid, ethyl ester (6CI, 8CI), Ethyl hexadecanoate, Ethyl palmitate, NSC 8918, Hexadecanoic acid ethyl ester, Palmitic acid ethyl ester
IUPAC Name ethyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C18H36O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20-4-2/h3-17H2,1-2H3
Formula Weight 284.2715
Chemical Name or Material Ethylpalmitate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.