missing translation for 'onlineSavingsMsg'
Learn More

Ethyl phenylpropiolate, 98+%

Product Code. 11310134
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11310134 25 g 25g
11300134 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11310134 Supplier Thermo Scientific Alfa Aesar Supplier No. L00603.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2216-94-6 | C11H10O2 | 174.199 g/mol

Ethyl phenylpropiolate, is an important organic intermediate. It is also used as an intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl phenylpropiolate, is an important organic intermediate. It is also used as an intermediate in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Heat sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2216-94-6
Molecular Formula C11H10O2
Molecular Weight (g/mol) 174.199
MDL Number MFCD00009185
InChI Key ACJOYTKWHPEIHW-UHFFFAOYSA-N
Synonym ethyl phenylpropiolate, ethylphenylpropiolate, ethyl 3-phenylpropiolate, ethyl phenylpropriolate, 2-propynoic acid, 3-phenyl-, ethyl ester, ethyl 3-phenylpropynoate, ethyl phenylpropynoate, ethyl phenylacetylenecarboxylate, phenylpropiolic acid ethyl ester, ethyl 3-phenyl-2-propynoate
PubChem CID 91516
IUPAC Name ethyl 3-phenylprop-2-ynoate
SMILES CCOC(=O)C#CC1=CC=CC=C1

Especificaciones

Density 1.055
Boiling Point 152°C to 153°C (21 mmHg)
Flash Point >110°C (230°F)
Odor Odorless
Refractive Index 1.552
Quantity 25 g
Beilstein 639637
Solubility Information Insoluble in water.
Formula Weight 174.2
Percent Purity ≥98%
Chemical Name or Material Ethyl phenylpropiolate
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.