Learn More
Ethyl propenyl ether, cis + trans, 97%
Product Code. 11461190
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11461190 100 g 100g
11441190 5 g 5g
11451190 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11461190 Supplier Thermo Scientific Alfa Aesar Supplier No. H26319.22
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 928-55-2 | C5H10O | 86.13 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 928-55-2
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00015184
InChI Key XDHOEHJVXXTEDV-HYXAFXHYSA-N
Synonym 1-ethoxypropene, ethyl propenyl ether, ether, ethyl propenyl, propenyl ethyl ether, e-1-ethoxyprop-1-ene, 1-ethoxy-1-propene, unii-jh817due12, ethyl 1-propenyl ether, 1-propene, 1-ethoxy, ethyl-1-propenyl ether
PubChem CID 5365091
SMILES CCO\C=C/C
Specifications
Density 0.778
Boiling Point 69°C to 71°C
Flash Point 20°C (68°F)
Refractive Index 1.398
Quantity 100 g
UN Number UN3271
Beilstein 1735974
IUPAC Name (E)-1-ethoxyprop-1-ene
Formula Weight 86.13
Percent Purity 97%
Chemical Name or Material Ethyl propenyl ether, cis + trans