missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Propionate(Propionic Acid Ethyl Ester), TRC

Product Code. 30409789
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409789 10 g -
30409113 1 g -
30419215 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30409789 Supplier Toronto Research Chemicals Supplier No. TRCP78636510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-37-3
Molecular Formula C5 H10 O2
Molecular Weight (g/mol) 102.13
Synonym Propionic acid, ethyl ester (6CI,8CI), Ethyl propanoate, Ethyl propionate, NSC 8848, Propionic ester, Propionic ether
IUPAC Name ethyl propanoate
SMILES CCOC(=O)CC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C5H10O2/c1-3-5(6)7-4-2/h3-4H2,1-2H3
Formula Weight 102.0681
Chemical Name or Material Propanoic acid ethyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.