missing translation for 'onlineSavingsMsg'
Learn More

Ethyl pyrazolo[1,5-a]pyridine-3-carboxylate, 98%, BLD Pharm™

Product Code. 30784545
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Unit Size:
1g
250mg
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30784545 5 g -
30781505 250 mg 250mg
30780032 1 g 1g
30781985 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30784545 Supplier BLD Pharm Supplier No. BD1295025g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 16205-44-0
Molecular Formula C10H10N2O2
MDL Number MFCD06411542
Quantity 5 g
SMILES O=C(C1=C2C=CC=CN2N=C1)OCC
Molecular Weight (g/mol) 190.20
Percent Purity 98%
Chemical Name or Material Ethyl pyrazolo[1,5-a]pyridine-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.