missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (R)-(+)-1-Boc-4-oxopiperidine-2-carboxylate, 95%, Thermo Scientific™

Product Code. 15423046
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15423046 1 g 1g
15433046 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15423046 Supplier Thermo Scientific Alfa Aesar Supplier No. H57019.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

It is used as pharmaceutical intermediate.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in chloroform and methanol.

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 357154-16-6
Molecular Formula C13H21NO5
Molecular Weight (g/mol) 271.313
MDL Number MFCD20526405
InChI Key YAVQLRUBKDCOCI-SNVBAGLBSA-N
Synonym r-1-tert-butyl 2-ethyl 4-oxopiperidine-1,2-dicarboxylate, ethyl r-1-boc-4-oxopiperidine-2-carboxylate, r-+-1-boc-4-oxopiperidine-2-carboxylic acid ethyl ester, r-1-boc-4-oxopiperidine-2-carboxylic acid ethyl ester, ethyl r-+-1-boc-4-oxopiperidine-2-carboxylate, ethyl r-+-1-boc-4-oxopipecolate, r-1-tert-butyl-2-ethyl-4-oxopiperidine-1,2-dicarboxylate, 2r-4-oxopiperidine-1,2-dicarboxylic acid 1-tert-butyl 2-ethyl ester, 1-tert-butyl 2-ethyl 2r-4-oxopiperidine-1,2-dicarboxylate
PubChem CID 67586303
IUPAC Name 1-O-tert-butyl 2-O-ethyl (2R)-4-oxopiperidine-1,2-dicarboxylate
SMILES CCOC(=O)C1CC(=O)CCN1C(=O)OC(C)(C)C

Specifications

Quantity 1 g
Solubility Information Soluble in chloroform and methanol.
Optical Rotation +20° (c=0.5 in Chloroform)
Formula Weight 271.31
Percent Purity 95%
Chemical Name or Material Ethyl (R)-(+)-1-Boc-4-oxopiperidine-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.