missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Tosylate, TRC

Product Code. 30377320
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30377320 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30377320 Supplier Toronto Research Chemicals Supplier No. TRCE92706510MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Tosylate
CAS 80-40-0
Purity Grade Notes HPLC
Molecular Formula C9 H12 O3 S
Quantity 10 mg
Synonym 4-Methylbenzenesulfonic Acid Ethyl Ester, p-Toluenesulfonic acid, ethyl ester (6CI,7CI,8CI), Ethyl 4-methylbenzenesulfonate, Ethyl 4-toluenesulfonate, Ethyl p-methylbenzenesulfonate, Ethyl p-toluenesulfonate, Ethyl p-tosylate, Ethyl toluenesulfonate, Ethyl tosylate, NSC 8887, Toluene-4-sulfonic acid ethyl ester
InChI Formula InChI=1S/C9H12O3S/c1-3-12-13(10,11)9-6-4-8(2)5-7-9/h4-7H,3H2,1-2H3
SMILES CCOS(=O)(=O)c1ccc(C)cc1
IUPAC Name ethyl 4-methylbenzenesulfonate
Molecular Weight (g/mol) 200.26
Formula Weight 200.051
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.