missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol ethyl methyl ether, 97%, stab. with 0.01% BHT, Thermo Scientific Chemicals

Product Code. p-7025379
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11495746

Brand: Thermo Scientific Alfa Aesar B20819.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is employed in the reaction of metal salts of thymine and uracil with tetra-acetyl-α-glucopyranosyl bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the reaction of metal salts of thymine and uracil with tetra-acetyl-α-glucopyranosyl bromide.

Solubility
Fully miscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5137-45-1
Molecular Formula C5H12O2
MDL Number MFCD00048580
Synonym 2,5-dioxaheptane, ethane, 1-ethoxy-2-methoxy, ethyl 2-methoxyethyl ether

Specifications

Density 0.85
Boiling Point 100°C to 102°C
Flash Point 2°C (36°F)
Linear Formula CH3OCH2CH2OCH2CH3
Refractive Index 1.387
Quantity 10 g
UN Number UN3271
Solubility Information Fully miscible in water.
Formula Weight 104.15
Percent Purity 97%
Chemical Name or Material Ethylene glycol ethyl methyl ether, stabilized with 0.01% BHT
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.