missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediamine-N,N,N’,N’-tetraacetic Acid-13C4, TRC

Product Code. 30136262
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136262 10 mg 10mg
30004037 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136262 Supplier Toronto Research Chemicals Supplier No. TRCE91774710MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1215408-17-5
Molecular Formula 13C4 C6 H16 N2 O8
Molecular Weight (g/mol) 296.2133
Synonym N,N’-1,2-Ethanediylbis[N-(carboxymethyl)glycine-13C4, Celon ATH-13C4, Cheelox-13C4, EDTA-13C4, Edathamil-13C4, Edetic Acid-13C4, Endrate-13C4, Ethylene-N,N'-biscarboxymethyl-N,N’-diglycine-13C4, Ethylenedinitrilotetraacetic Acid-13C4, Sequestric Acid-13C4, Sequestrol-13C4, Titriplex-13C4, Trilon BW-13C4, Trilon BX-13C4, Versene-13C4;
IUPAC Name 2-[2-[bis(hydroxycarbonylmethyl)amino]ethyl-(hydroxycarbonylmethyl)amino]acetic acid
SMILES O[13C](=O)CN(CCN(C[13C](=O)O)C[13C](=O)O)C[13C](=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H16N2O8/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20)/i7+1,8+1,9+1,10+1
Formula Weight 296.1041
Chemical Name or Material Ethylenediamine-N,N,N’,N’-tetraacetic Acid-13C4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.