missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediamine-N,N,N',N'-tetraacetic Acid Disodium Dihydrate, TRC

Product Code. 30010554
Change view
Click to view available options
Quantity:
2.5 g
10 g
500 g
Unit Size:
10g
2.5g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010554 500 g 500g
30007986 10 g 10g
17944695 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010554 Supplier Toronto Research Chemicals Supplier No. TRCE917735500G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H14N2O8.2Na.2H2O
Molecular Weight (g/mol) 372.24
Synonym Disodium edetate, Disodium dihydrogen (ethylenedinitrilo)tetraacetate dihydrate, EDTA Disodium Dihydrate
IUPAC Name disodium;2-[2-[carboxylatomethyl(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetate;dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(=O)O)CC(=O)[O-])CC(=O)[O-]

Specifications

Quantity 500 g
InChI Formula InChI=1S/C10H16N2O8.2Na.2H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;2*1H2/q;2*+1;;/p-2
Formula Weight 372.0757
Percent Purity >95%
Chemical Name or Material Disodium edetate dihydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.