missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ethylenediaminetetraacetic acid trisodium salt hydrate, 95%

Product Code. 15472516
Change view
Click to view available options
Quantity:
250 g
500 g
Unit Size:
250g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15472516 500 g 500g
15462516 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15472516 Supplier Alfa Aesar™ Supplier No. H56165.36

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
This is used to eliminate enzyme inhibition by traces of heavy metals, and to inhibit enzymes that require divalent cations as cofactors.

Solubility
Soluble in sodium chloride. (1 M NaOH).

Notes
Incompatible with oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85715-60-2
Molecular Formula C10H15N2Na3O9
Molecular Weight (g/mol) 376.204
MDL Number MFCD00149675
InChI Key KKTNQJMZBPLVKM-UHFFFAOYSA-K
Synonym trisodium hydrogen ethylenediaminetetraacetate hydrate, tricon bw, unii-zmn5a047nl, endrate, edta triasodium salt, zmn5a047nl, ethylenediaminetetraacetic acid trisodium salt hydrate, sequestrene sodium 3, trisodium edta monohydrate, edetate trisodium monohydrate
PubChem CID 16211179
IUPAC Name trisodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxymethyl)amino]acetate;hydrate
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)[O-].O.[Na+].[Na+].[Na+]

Specifications

Melting Point >300°C
Quantity 500 g
Formula Weight 358.19
Percent Purity 95%
Chemical Name or Material Ethylenediaminetetraacetic acid trisodium salt hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.