missing translation for 'onlineSavingsMsg'
Learn More

Ethyltriphenylphosphonium bromide, 98+%

Product Code. 11406296
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11406296 25 g 25g
11416296 100 g 100g
11426296 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11406296 Supplier Thermo Scientific Alfa Aesar Supplier No. B23096.14
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1530-32-1 | C20H20BrP | 371.26 g/mol

Ethyltriphenylphosphonium bromide acts as a reactant in the synthesis of D-amino acids from L-cysteine-derived thiazolidines, Leiodolide A through aldol reactions and Horner-Wadsworth-Emmons olefination. It is also used in the preparation of cycloalkanoindolines through diastereoselective intramolecular inimo-ene reactions. Further, it is used as a reagent in solid-state metathesis polycondensation to prepare alkyl-dipropenylthiophene monomers and Mizoroki-Heck cyclization and cascading Tsuji-Trost cyclization reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyltriphenylphosphonium bromide acts as a reactant in the synthesis of D-amino acids from L-cysteine-derived thiazolidines, Leiodolide A through aldol reactions and Horner-Wadsworth-Emmons olefination. It is also used in the preparation of cycloalkanoindolines through diastereoselective intramolecular inimo-ene reactions. Further, it is used as a reagent in solid-state metathesis polycondensation to prepare alkyl-dipropenylthiophene monomers and Mizoroki-Heck cyclization and cascading Tsuji-Trost cyclization reactions.

Solubility
Soluble in methanol. Slightly soluble in water, acetone and isopropanol.

Notes
Hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1530-32-1
Molecular Formula C20H20BrP
Molecular Weight (g/mol) 371.26
MDL Number MFCD00011838
InChI Key JHYNXXDQQHTCHJ-UHFFFAOYSA-M
Synonym ethyltriphenylphosphonium bromide, ethyl triphenyl phosphonium bromide, triphenylethylphosphonium bromide, phosphonium, ethyltriphenyl-, bromide, ethyltriphenylphosphanium bromide, ethyl triphenylphosphonium bromide, unii-r85v84ul5v, ethyltriphenylphosphoniumbromide, guaifenesin powder, pubchem18705
PubChem CID 73727
SMILES [Br-].CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifikationer

Melting Point 206°C to 210°C
Flash Point >200°C (392°F)
Odor Odorless
Linear Formula CH3CH2P(C6H5)3Br
Quantity 25 g
UN Number UN3077
Beilstein 3599630
Sensitivity Hygroscopic
Solubility Information Soluble in methanol. Slightly soluble in water,acetone and isopropanol.
IUPAC Name ethyl(triphenyl)phosphanium;bromide
Formula Weight 371.26
Percent Purity ≥98%
Chemical Name or Material Ethyltriphenylphosphonium bromide
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.