missing translation for 'onlineSavingsMsg'
Learn More

Etidronate, TRC

Product Code. 30382826
Change view
Click to view available options
Quantity:
50 mg
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382826 50 mg
30387897 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30382826 Supplier Toronto Research Chemicals Supplier No. TRCE93499550MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2809-21-4
Molecular Formula C2 H8 O7 P2
Molecular Weight (g/mol) 206.028
Synonym Phosphonic acid, P,P'-(1-hydroxyethylidene)bis-, Phosphonic acid, (1-hydroxyethylidene)bis- (9CI), Phosphonic acid, (1-hydroxyethylidene)di- (8CI), P,P'-(1-Hydroxyethylidene)bis[phosphonic acid\], (1-Hydroxyethylidene)-1,1-bis(phosphonic acid), (1-Hydroxyethylidene)-1,1-diphosphonic acid, (1-Hydroxyethylidene)bisphosphonic acid, (1-Hydroxyethylidene)diphosphonic acid, 1-Hydroxy-1,1-diphosphonoethane, 1-Hydroxyethane-1,1-bisphosphonic acid, 1-Hydroxyethane-1,1-diphosphonic acid, 1-Hydroxyethane-1,1-diyldiphosphonic acid, 1-Hydroxyethanediphosphonic acid, 1-Hydroxyethylidene-1,1-diphoshonic acid, 1-Hydroxyethylidene-1,1-diphosphoric acid, 1-Hydroxyethylidene-1,1'-diphosphonic acid, 1000SL, Acetodiphosphonic acid, Acidum etidronicum, Anti Cal 5, Aquacid 105, Aquacid 105CG, Aquacid 105EX, Aquacid 105EXLC, Aquacid 105EXSP, Aquacid 105NS, Belclene 660, Briquest ADPA 60A, Briquest ADPA-A, Chelate 101, Chelest PH 210L, Cublen K 60, Defloc EH 06, Dequest 16, Dequest 2010, Dequest 2010CS, Dequest 2010LA, Dequest 2010R, Dequest 2015, Dequest D 2010, Dequest Z 010, EHDP, Ethane-1-hydroxy-1,1-diphosphonic acid, Ethidronate, Ethydronate, Etidronate, Etidronic acid, Feliox 115, Feliox 115A, Feliox CY 115, Ferrofos 510, H 501, H 501 (corrosion inhibitor), HDEPA, HEDP, Hydroxyethanediphosphonic acid, Kochtreat 75, Ksidifon, Lonza 106, Mascol P 210, Masquol P 210, Mayoquest 1500, NSC 227995, OEDF, OEDFK, OEDP, Oxyethylidenediphosphonic acid, RP 61, Sequion 10H, Sone 16, Tecquest 360, Terpil SL, Turpinal SL, Turpinal SLR, Unihib 106, Wayplex, Xidifon, Xidiphone, Xydiphone
IUPAC Name (1-hydroxy-1-phosphonoethyl)phosphonic acid
SMILES CC(O)(P(=O)(O)O)P(=O)(O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C2H8O7P2/c1-2(3,10(4,5)6)11(7,8)9/h3H,1H3,(H2,4,5,6)(H2,7,8,9)
Formula Weight 205.975
Chemical Name or Material Etidronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.