Description
IPGphor Strip Holder Cleaning Solution has been specifically formulated for removing protein deposits. Strip Holder Cleaning Solution is supplied in 1L bottle.
Compatible with these stripholders and trays used with Immobiline™ DryStrips:
- IPGphor Manifold Ceramic Tray
- IPGphor strip holders (various sizes)
- IPGphor Cup Loading Manifold
- IPGphor Manifold Light
Specifications
Specifications
| For Use With (Application) | Designed for mild cleaning of Ceramic Ettan™ IPGphor™ strip holders, manifold Ceramic tray, cup loading manifold and manifold light used for isoelectric focusing of Protein samples |
| Volume (Metric) | 1 L |