Learn More
Cytiva Ettan™ IPGphor™ Strip Holder Cleaning Solution
Product Code. 12623486 Shop All Cytiva Products
Change view
Click to view available options
:
IPGphor Strip Holder Cleaning Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12623486 IPGphor Strip Holder Cleaning Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12623486 Supplier Cytiva Supplier No. 29011927
Strip Holder Cleaning Solution is designed for mild cleaning of ceramic stripholders used for isoelectric focusing of protein samples
IPGphor Strip Holder Cleaning Solution has been specifically formulated for removing protein deposits. Strip Holder Cleaning Solution is supplied in 1L bottle.
Compatible with these stripholders and trays used with Immobiline™ DryStrips:
  • IPGphor Manifold Ceramic Tray
  • IPGphor strip holders (various sizes)
  • IPGphor Cup Loading Manifold
  • IPGphor Manifold Light
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Designed for mild cleaning of Ceramic Ettan™ IPGphor™ strip holders, manifold Ceramic tray, cup loading manifold and manifold light used for isoelectric focusing of Protein samples
Volume (Metric) 1 L