Learn More
Corning™ Euro Adaptor
Product Code. 10758098 Shop All Corning Products
Change view
Click to view available options
Voltage: 15VAC
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10758098 Voltage: 15VAC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10758098 Supplier Corning™ Supplier No. 4915A
  • ONLINE EXCLUSIVE PRICE
    83.45€
    77.96€ / Pack of 1
    Save 5.49€
    6% Off
Estimated Shipment: 07-12-2026
Add to basket
Add to basket
Recharger for Corning™ Stripettor™ pipette
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Corning™ Stripettro™ Pipette