Learn More
Thermo Scientific™ Eutech™ USB to IrDA Interface Adapter for Eutech™ 600 Series meters
Product Code. 11583835
Change view
Click to view available options
For Use With (Equipment):
600 / 610 / 620 / 650 meters
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11583835 600 / 610 / 620 / 650 meters Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11583835 Supplier Thermo Scientific™ Supplier No. 01X447602
  • ONLINE EXCLUSIVE PRICE
    179.00€
    140.40€ / Pack of 1
    Save 38.60€
    21% Off
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Eutech™ offers various types of adapters that are available for use with different meters for PC communication.
Eutech™ offers various types of adapters that are available for use with different meters for PC communication. The Eutech™ USB to IrDA Interface Adapter is used to connect Eutech™ 600 Series meters to your computer.
Features:
  • Used With: Eutech 600 / 610 / 620 / 650
  • Note: This product is not compatible with Windows 10 and 11
TRUSTED_SUSTAINABILITY
Specifications
Description USB / IrDA Interface Adapter (for 600 Series Meter to PC Communication)
Type USB to IrDA Interface Adapter
Compatibility Not compatible with Windows 10 and 11
For Use With (Equipment) 600 / 610 / 620 / 650 meters