missing translation for 'onlineSavingsMsg'
Learn More

Evans blue (Technical Grade), TRC

Product Code. 30380342
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30380342 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380342 Supplier Toronto Research Chemicals Supplier No. TRCE94520310G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Evans blue (Technical Grade)
CAS 314-13-6
Molecular Formula C34H24N6Na4O14S4
Quantity 10 g
Synonym Evans Blue tetrasodium salt
InChI Formula InChI=1S/C34H28N6O14S4.4Na/c1-15-11-17(3-7-21(15)37-39-23-9-5-19-25(55(43,44)45)13-27(57(49,50)51)31(35)29(19)33(23)41)18-4-8-22(16(2)12-18)38-40-24-10-6-20-26(56(46,47)48)14-28(58(52,53)54)32(36)30(20)34(24)42;;;;/h3-14,41-42H,35-36H2,1-2H3,(H,43,44,45)(H,46,47,48)(H,49,50,51)(H,52,53,54);;;;/q;4*+1/p-4
SMILES [Na+].[Na+].[Na+].[Na+].Cc1cc(ccc1N=Nc2ccc3c(cc(c(N)c3c2O)S(=O)(=O)[O-])S(=O)(=O)[O-])c4ccc(N=Nc5ccc6c(cc(c(N)c6c5O)S(=O)(=O)[O-])S(=O)(=O)[O-])c(C)c4
IUPAC Name tetrasodium;4-amino-6-[(E)-[4-[4-[(E)-(8-amino-1-hydroxy-5,7-disulfonatonaphthalen-2-yl)diazenyl]-3-methylphenyl]-2-methylphenyl]diazenyl]-5-hydroxynaphthalene-1,3-disulfonate
Molecular Weight (g/mol) 960.81
Formula Weight 959.9824
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.