Learn More
Lenz Laborglasintrumente™ Evaporating Flasks with Conical Ground Joint
Product Code. 11923391
Change view
Click to view available options
Volume (Metric):
500 mL
1000 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11923391 500 mL Each
11933391 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11923391 Supplier Lenz Laborglasintrumente™ Supplier No. 03044058
  • ONLINE EXCLUSIVE PRICE
    140.00€
    98.43€ / Pack of 1
    Save 41.57€
    29% Off
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
DURAN™ glass evaporating flask with highly transparent PUR coating
  • Especially for rotary evaporators
  • With center neck and conical joint
  • With highly transparent PUR coating
  • Autoclavable
  • Pear-shaped
  • Material: DURAN™
Specifications
Material Duran Glass
For Use With (Equipment) Rotary Evaporators
Material (Coating) PUR
Quantity 1
Shape Pear
Volume (Metric) 500 mL
Diameter (Metric) 101 mm