Learn More
Lenz Laborglasintrumente Evaporating Flasks with Conical Ground Joint, with PUR Coating
Product Code. 11993381
Change view
Click to view available options
Volume (Metric):
50 mL
100 mL
250 mL
2 L
3 L
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11993381 50 mL Each
11903391 100 mL Each
15695585 250 mL Each
11943391 2 L Each
11953391 3 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11993381 Supplier Lenz Laborglasintrumente Supplier No. 03044028
  • ONLINE EXCLUSIVE PRICE
    94.00€
    61.42€ / Pack of 1
    Save 32.58€
    34% Off
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Especially for rotary evaporators
  • With center neck and conical joint
  • With highly transparent PUR coating
  • Autoclavable
  • Pear-shaped
  • Material: DURAN™
Specifications
Material Duran Glass
Volume (Metric) 50 mL
Diameter (Metric) 56 mm