missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EVI2B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99056-100ul
This item is not returnable.
View return policy
Description
EVI2B Polyclonal antibody specifically detects EVI2B in Mouse samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| EVI2B | |
| Polyclonal | |
| Western Blot 1:500-1:1000, Immunoprecipitation 0.2-1 uL/mg of lysate | |
| CD361 antigen, D17S376, ecotropic viral integration site 2B, Ecotropic viral integration site 2B protein homolog, EVDBprotein EVI2B, EVI-2B | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Mouse EVI2B. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 2124 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction