Learn More
Thermo Scientific™ eVol™ XR Dispensing System Charger and Stand
Product Code. 10338279 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Dispensing System Charger
Dispensing System Stand
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
10338279 Dispensing System Charger Each
12504356 Dispensing System Stand Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10338279 Supplier Thermo Scientific™ Supplier No. 66002027
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Recharge your Thermo Scientific™ eVol™ Dispensing System with this charger, and get the stand that holds your eVol Dispensing System when not in use.
Order the Thermo Scientific™ eVol™ Dispensing System Charger and Stand to recharge your dispensing system. The eVol dispenser is a unique, hand-held digital dispenser that accurately and reproducibly performs a wide variety of liquid handling procedures. The easy-to-use device couples an analytical syringe to a programmable electronic controller, enabling highly reproducible dispensing for all users.
The eVol Dispensing System is ideal for laboratories that need to repeatedly dispense nonaqueous liquids, offering greater accuracy and reproducibility for applications such as:
  • Routine dispensing of volatile solvents, hazardous, corrosive or viscous chemicals
  • Preparation of calibration standards
  • Addition of internal standards
The system unifies two precision devices: a digitally controlled electronic drive, and a Thermo Scientific™ XCHANGE™ enabled analytical syringe.
Repeatable Performance for Higher Confidence in Your Results
  • Can be calibrated easily by a user to ensure validity of results
  • Intuitive, easy-to-use interface with touch wheel and full-color screen
  • Complies with GLP and GMP guidelines
Exceptional Versatility and Functionality
  • Integrated XCHANGE coupling allows syringes to be quickly and easily changed, preventing possible cross-contamination
  • Addition of Thermo Scientific™ MEPS™ (Micro Extraction by Packed Sorbent) technology to an XCHANGE syringe enables automated sample preparation by SPE in a hand-held system
TRUSTED_SUSTAINABILITY
Specifications
Product Type Dispensing System Charger
Product Line HyperSep
For Use With (Equipment) Electronic Dispensing System