Learn More
Safety First Aid Evolution HypaSoothe Burns Kit, Large
Product Code. 31759410
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Unit Size
31759410 Sterile Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31759410 Supplier Safety First Aid Supplier No. K575
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The kit is a specialised first aid kit designed specifically for the treatment of minor burn injuries. It contains fast acting quick-relief dressings and gels to stop the burn process and soothe injuries.
  • All kits include HypaSoothe burn dressings which are sterile gel impregnated dressings.
  • Gel soaked burn dressings help prevent contamination.
  • For use on 1st and 2nd degree burns.
  • Cools the burn and sooths pain, Pre cooling not required (as per BS 8599 standard).
  • The Evolution HypaSoothe Burns Kit, Large, Contains:
    • 1x First Aid Guidance Leaflet
    • 2x HypaSoothe Burn Dressings, 10 x 10 cm
    • 2x HypaSoothe Burn Dressing, 20 x 20 cm
    • 9x HypaSoothe Burn Gel Sachets, 6 g
    • 12x HypaBand Safety Pins
    • 4x HypaBand Conforming Bandages
    • 1x Snips Clothing Cutters, 15 cm
    • 4x HypaTouch Disposable Gloves (Pair)
Specifications
Product Type First Aid Kit
Material Plastic
Sterility Sterile
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.