Learn More
Safety First Aid Evolution Plus Eyewash Kit with Mirror and 2 x 500 mL Eyewash Bottles
Descripción
- The kit is supplied in a durable hinged case that includes bottle stabilisers to hold contents firmly in place.
- It includes a quick release wall fixing bracket and kit identification label to provide a better storage and an easier identification.
- Once on the bracket, the kit can be opened and worked with, providing a self-supporting workbench.
- Evolution Plus Eyewash Kit assures you have adequate eye wash facilities at your place of work and is available for purchase, with or without mirror.
- Evolution Plus Eye Wash with Mirror Contains:
- 2x HypaClens Eye Wash Bottles, 500 mL
- 2x HypaCover Sterile Eye Dressings
Especificaciones
Especificaciones
| Color | Clear |
| Flow Rate | 20 gpm (Shower), 8.0 gpm (Eye and Face Wash) |
| Material | Plastic |
| Mounting | Wall |
| Product Type | Portable Eyewash and Shower Fixtures |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?