Learn More
Invitrogen™ EVOS™ Light Cube, CFP 2.0
Artikelnummer. 17324493
Ansicht ändern
Click to view available options
Quantity:
1 each
Packungsgröße:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packungsgröße
17324493 1 each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17324493 Lieferant Invitrogen™ Lieferanten-Nr. AMEP4953
Black Friday Starts Here
Use promo code 27984
Alle Werbeaktionen anzeigen
 Restricted Item
Estimated Shipment: 11-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
EVOS LED light cubes provide superior illumination uniformity across the field of view, resulting in exceptional tile-stitch image quality. At the same time, optimized spectral fidelity across channels significantly improves data quality by minimizing undesired bleed-through.
  • Second-generation EVOS LED light cubes provide superior illumination uniformity across the field of view, resulting in exceptional tile-stitch image quality
  • At the same time, optimized spectral fidelity across channels significantly improves data quality by minimizing undesired bleed-through
  • Like the original design, they combine condensing optics and hard-coated filters into one light engine that delivers digitally controlled fluorophore excitation
  • These LED light cubes are backwards compatible with older EVOS and Countess systems. and can be used with first-generation light cubes
  • Like the original light cubes, these LED cubes are user-changeable and automatically recognized by the instrument
  • The EVOS Light Cube, CFP 2.0, is ideal for use with CellLight CFP, ECFP, Evans Blue, Lucifer Yellow, and other fluorophores with excitation and emission maxima near 445 and 510 nm
  • The LED bulbs on the EVOS systems are rated for >50,000 hours, compared to 300 hours for a typical mercury bulb and 1,500 hours for a metal halide bulb
EVOS LED light cubes offer these important advantages:
  • Uniform illumination across the entire field of view for superb image quality
  • Optimized spectral compatibility to maximize true signal detection, while minimizing bleed-through for the best possible data quality
  • A short light path that affords superior detection of fluorescent signals
  • Stable illumination intensity over 50,000 hours that helps experimental reproducibility
TRUSTED_SUSTAINABILITY
Spezifikation
Type LED Light Cube
Product Line EVOS
For Use With (Equipment) Countess™ III FL Automated Cell Counter, EVOS™ M5000 Imaging System, EVOS™ M7000 Imaging System
Includes Condensing Optics, Hard-coated Filters
Wavelength Ex: 445/45 Em: 510/42
Quantity 1 each
For Research Use Only. Not for use in diagnostic procedures.
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.