missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EVPL Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18402C
This item is not returnable.
View return policy
Description
EVPL Polyclonal antibody specifically detects EVPL in Human samples. It is validated for Western Blot
Specifications
| EVPL | |
| Polyclonal | |
| Western Blot | |
| 210 KDa Cornified Envelope Precursor Protein, 210 KDa Paraneoplastic Pemphigus Antigen, envoplakin, EVPK, p210 | |
| Recognizes region between residues Between Between 1275 and 1325.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 2125 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction