missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals Exendin-4 Antibody (20), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-23577H
This item is not returnable.
View return policy
Description
Exendin-4 Monoclonal antibody specifically detects Exendin-4 in Reptile samples. It is validated for ELISA
Specifications
| Exendin-4 | |
| Monoclonal | |
| HRP | |
| Exendin4 | |
| Synthetic exendin-4 | |
| 0.1 mL | |
| Biologically Active Proteins | |
| Reptile | |
| Purified |
| ELISA | |
| 20 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction