Learn More
asecos™ Exhaust Air Unit
Product Code. 15957826 Shop All asecos Products
Change view
Click to view available options
For Use With (Equipment):
Tall cabinets
Under bench cabinets
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15957826 Under bench cabinets Each
13463877 Tall cabinets Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is under eco-contribution tax.
Product Code. 15957826 Supplier asecos™ Supplier No. 24732
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Extraction module for wall mounting, tall cabinets, or under bench cabinets with exhaust air monitoring
  • Extraction units/modules for installation on the cabinet or wall mounting – ready to plug-in, easy and quick installation
  • Safe capturing of escaping vapours and gases at the point where they are released or produced
  • Very low noise, ideal for direct installation at the workplace
  • With connection cable and power cord
  • Connecting fitting DN 75
  • ATEX compliant: C' II 3/-G Ex c llC T4
  • With extraction air monitoring
    • Integrated pressure leak detector
    • The green LED turns off in case of failure and the red LED lights up
Specifications
Certifications/Compliance CE compliant
Depth (Metric) Exterior 200 mm
Description For wall mounting or under bench cabinets, with exhaust air monitoring and with potential-free alarm contact, CE compliant: C ` II 3/- G Ex c IIC T4, explosion-proof, extraction from: EX-Zone 2 / installation: outside any EX-ZONE
Dimensions (D x W x H) 200 x 400 x 340 mm
For Use With (Application) Extraction module for wall mounting – ready to plug-in, easy and quick installation
For Use With (Equipment) Under bench cabinets
Height (Metric) Exterior 340 mm
Includes Installation bracket, flexible hose, hose clips and installation material
Model HF.EA.24139
Surface Coating Powder-coated plain
Weight (Metric) 12 kg
Width (Metric) Exterior 400 mm
Material Steel
Product Type Exhaust air unit