missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Exosome component 5 Antibody (V80P1C10*C) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
190.00€ - 351.00€
Specifications
| Antigen | Exosome component 5 |
|---|---|
| Clone | V80P1C10*C |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18648088
|
Novus Biologicals
NBP2-50173-0.025ml |
0.025 mL |
190.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18640517
|
Novus Biologicals
NBP2-50173 |
0.1 mL |
351.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Exosome component 5 Monoclonal antibody specifically detects Exosome component 5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunohistochemistrySpecifications
| Exosome component 5 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| DNA replication Transcription Translation and Splicing, Epigenetics | |
| PBS | |
| 56915 | |
| IgG | |
| Protein A purified |
| V80P1C10*C | |
| Western Blot 1:100 - 1:2000, Flow Cytometry, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CML28, exosome complex exonuclease RRP46, exosome component 5Chronic myelogenous leukemia tumor antigen 28, exosome component Rrp46, hRrp46p, MGC12901, p12BMGC111224, RRP41B, Rrp46p, RRP46Ribosomal RNA-processing protein 46 | |
| Peptide Sequence - KLLMSSTKGLY | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title