Learn More
Novus Biologicals™ Exosome Standards (Human Urine)
Product Code. 18654310 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 x 100 μg Vials
2 x 30 μg Vials
6 x 100 μg Vials
6 x 30 μg Vials
Unit Size:
Pack of 2
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18654310 2 x 30 μg Vials Pack of 2
18638420 6 x 100 μg Vials Pack of 6
18635370 2 x 100 μg Vials Pack of 2
18692640 6 x 30 μg Vials Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 18654310 Supplier Novus Biologicals™ Supplier No. NBP24984060ug
  • ONLINE EXCLUSIVE PRICE
    441.00€
    416.85€ / Pack of 2
    Save 24.15€
    5% Off
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at 4°C. After reconstitution store at -70°C.
Concentration LYOPH
Quantity 2 x 30 μg Vials