Learn More
R&D Systems™ Exosome Standards (LnCAP cell line)
Product Code. 18687640 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 x 100 μg Vials
2 x 30 μg Vials
6 x 100 μg Vials
6 x 30 μg Vials
Unit Size:
Pack of 2
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18687640 2 x 30 μg Vials Pack of 2
18657910 6 x 100 μg Vials Pack of 6
18620941 2 x 100 μg Vials Pack of 2
18652031 6 x 30 μg Vials Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 18687640 Supplier R&D Systems™ Supplier No. NBP31168760ug
  • ONLINE EXCLUSIVE PRICE
    472.36€
    416.85€ / Pack of 2
    Save 55.51€
    11% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A Novus product, part of R&D Systems. Highly pure, lyophilized exosome standards with superior stability, optimal for multiple applications
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at 4°C. After reconstitution store at -70°C.
Concentration LYOPH
Product Line Novus
Quantity 2 x 30 μg Vials