Learn More
Kartell SpA Extension Handle for Automatic Pipette Washer
Product Code. 10102621
Change view
Click to view available options
For Use With:
Pipet Washers
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Unit Size
10102621 Pipet Washers Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10102621 Supplier Kartell SpA Supplier No. 0121900
  • ONLINE EXCLUSIVE PRICE
    18.40€
    16.63€ / Pack of 2
    Save 1.77€
    9% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Extension handle for basket of item 0021900
Component of automatic pipette washer item code 0021900. Extension handle to be used for burettes
Specifications
For Use With Pipet Washers
Height (English) 11.53 in.
Height (Metric) 292.9 mm
Product Type Extension Handle