Learn More
BUCHI B-811 Extraction Chamber LSV
Product Code. 11957258 Shop All Buchi Products
Change view
Click to view available options
Capacity (Metric):
340 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11957258 340 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11957258 Supplier BUCHI Supplier No. 037902
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Extraction chamber with thread, LSV Borosilicate glass, L 289 mm
  • Threaded top for B-811
  • Large Sample Volume (LSV)
  • Borosilicate glass
  • Length: 289 mm
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) B811 Extraction System
Product Type Extraction Chamber
Capacity (Metric) 340 mL
For Use With (Application) Soxhlet extraction
Quantity 1 Pack