Learn More
Esculape Laboratoire™ Eyewash Station
Product Code. 11987625
Change view
Click to view available options
Includes:
0.9% Sodium Chloride, 2 x 500 mL
0.9% Sodium Chloride, 500 mL
200 mL Blue Neutral pH Solution, 500 mL Bottle Green Sterile Ophthalmic Solution
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
11987625 200 mL Blue Neutral pH Solution, 500 mL Bottle Green Sterile Ophthalmic Solution Each
10617204 0.9% Sodium Chloride, 2 x 500 mL Each
11769522 0.9% Sodium Chloride, 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11987625 Supplier Esculape Laboratoire™ Supplier No. 8205203
  • ONLINE EXCLUSIVE PRICE
    129.15€
    69.97€ / Pack of 1
    Save 59.18€
    45% Off
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Administer emergency eye care using an Esculape™ Eyewash Station. Components of this station include those that rinse and evacuate foreign substances from the eyes.
  • Fixed position with wall bracket;
  • Ergonimic eye cup aids continuous rinsing
  • Quick release bottle caps
  • Instructions in three languages
  • Three year shelf life if unopened
Specifications
For Use With (Application) For Remove Particles or Dust, Rehydrates the Eye
Includes 200 mL Blue Neutral pH Solution, 500 mL Bottle Green Sterile Ophthalmic Solution
Product Type Eyewash Station
Product Title
Select an issue
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.