Learn More
Thermo Scientific™ EZ-Link™ NHS-Biotin
Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Thermo Scientific™ EZ-Link NHS-Biotin is the simplest NHS-ester activated biotinylation reagent for labeling primary amines (e.g., protein lysines), whose membrane permeability enables it to be used for general intracellular labeling.
Highlights:
- Forms stable amide bonds
- Reacts with primary amines at pH 7-9
- Must be dissolved in DMSO or DMF and can then be transferred to an aqueous reaction solution
- Contains a shorter spacer arm than NHS-LC-Biotin
- Penetrates cell membranes because it is uncharged
Specifications
Specifications
| Product Type | NHS-Biotin |
| Quantity | 100 mg |
| Content And Storage | Store desiccated at room temperature. |
| Product Line | EZ-Link™ |
| Cell Permeability | Cell-Permeant |
| Chemical Reactivity | Amine |
| Reactive Moiety | Active Ester, Succinimidyl Ester, NHS Ester |
| Solubility | DMF (Dimethylformamide), DMSO (Dimethylsulfoxide) |
| Label or Dye | Biotin |
| Spacer | Short |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion