Learn More
Pacific Handy Cutter EZAR Auto-Retracting Lightweight Safety Cutter
Product Code. 17748977
Change view
Click to view available options
Color:
Blue
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17748977 Blue Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17748977 Supplier Pacific Handy Cutter Supplier No. EZAR
  • ONLINE EXCLUSIVE PRICE
    155.49€
    124.70€ / Pack of 12
    Save 30.79€
    19% Off
Estimated Shipment: 22-12-2026
Add to basket
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
The EZAR is a safety knife with fully automatic blade retraction. Therefore, it is particularly safe: the blade retracts fully automatically into the handle body, even if you leave your thumb on the slider.
  • In addition, it is equipped with a blade with a rounded tip to reduce the risk of puncturing.
  • The EZAR has a stable plastic body and is therefore particularly economical and lightweight.
  • Thanks to the three-sided slider, it can be used flexibly for horizontal and vertical cuts. And of course it can be used by both right- and left-handers.
  • The EZAR is very easy to open to clean it or change the blade.
  • The blade can be turned, which further increases the efficiency of the EZAR.
  • The metal tape splitter on the knife head additionally minimises use of your blade.
  • Especially suitable for thicker cutting materials, corrugated cardboard, strapping, adhesive tape, foil, textiles, felt, PVC, leather, rubber, string, etc.
Spezifikation
Blade Material Plastic
Product Type Auto Retracting Lightweight Safety Cutter
Disposable Reusable
Sterility Non-sterile
Color Blue
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.