missing translation for 'onlineSavingsMsg'
Learn More

Ezetimibe Ketone, Mikromol

Product Code. 30457572
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457572 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457572 Supplier Mikromol Supplier No. MM0935.010025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Ezetimibe Ketone, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ezetimibe Ketone
CAS 191330-56-0
Analyte or Component Names Ezetimibe Ketone
Shipping Condition Room Temperature
Molecular Formula C24 H19 F2 N O3
Certifications/Compliance ISO 9001
Synonym 2-Azetidinone, 1-(4-fluorophenyl)-3-[3-(4-fluorophenyl)-3-oxopropyl]-4-(4-hydroxyphenyl)-, (3R,4S)-, 2-Azetidinone, 1-(4-fluorophenyl)-3-[3-(4-fluorophenyl)-3-oxopropyl]-4-(4-hydroxyphenyl)-, (3R-trans)-, (3R,4S)-1-(4-Fluorophenyl)-3-[3-(4-fluorophenyl)-3-oxopropyl]-4-(4-hydroxyphenyl)azetidin-2-one
InChI Formula InChI=1S/C24H19F2NO3/c25-17-5-1-15(2-6-17)22(29)14-13-21-23(16-3-11-20(28)12-4-16)27(24(21)30)19-9-7-18(26)8-10-19/h1-12,21,23,28H,13-14H2/t21-,23-/m1/s1
Solution Type Neat
SMILES Oc1ccc(cc1)[C@@H]2[C@@H](CCC(=O)c3ccc(F)cc3)C(=O)N2c4ccc(F)cc4
IUPAC Name (3R,4S)-1-(4-fluorophenyl)-3-[3-(4-fluorophenyl)-3-oxopropyl]-4-(4-hydroxyphenyl)azetidin-2-one
Molecular Weight (g/mol) 407.41
Quantity 25 mg
Formula Weight 407.1333 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.