missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F4/80 Antibody (CI-A3-1) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 64 publications
Brand: Novus Biologicals NB600-404
This item is not returnable.
View return policy
Description
F4/80 Monoclonal specifically detects F4/80 in Human, Mouse, C. elegans, E. coli samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Radioimmunoassay, Electron Microscopy.
Specifications
| F4/80 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500RadioImmunoassay 1:100-1:2000, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Radioimmunoassay 1:100-1:2000, Electron Microscopy 1:10-1:500 | |
| ADGRE1, Adhesion G protein-coupled receptor E1, cell surface glycoprotein F4/80, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, EMR1 hormone receptor, F4/80, Gpf480, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7LN3 | |
| Rat | |
| 160 kDa | |
| 0.125 mg | |
| Immune System Diseases, Immunology, Innate Immunity, Myeloid derived Suppressor Cell | |
| 2015 | |
| Human, Mouse, C. elegans, E. coli | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| CI-A3-1 | |
| Unconjugated | |
| Q61549 | |
| ADGRE1 | |
| Thioglycollate stimulated peritoneal macrophages from C57BL/6 mice. | |
| Protein G purified | |
| RUO | |
| Primary | |
| This recognizes mouse F4/80, a 160kD glycoprotein expressed by murine macrophages. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction