missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ F4/80 Monoclonal Antibody (BM8), Brilliant Ultra Violet™ 805, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
191.00€ - 387.00€
Specifications
| Antigen | F4/80 |
|---|---|
| Clone | BM8 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30245383
|
Invitrogen™
368-4801-80 |
25 μg |
191.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30245046
![]() |
Invitrogen™
368-4801-82 |
100 μg |
387.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
F4/80 Monoclonal antibody specifically detects F4/80 in Mouse samples. It is validated for Flow CytometrySpecifications
| F4/80 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| ADGRE1; Adhesion G protein-coupled receptor E1; Adhesion G protein-coupled receptor E2; cell surface glycoprotein EMR1; cell surface glycoprotein F4/80; DD7A5-7; EGF-like module containing, mucin-like, hormone receptor-like 1; EGF-like module containing, mucin-like, hormone receptor-like sequence 1; EGF-like module receptor 1; EGF-like module-containing mucin-like hormone receptor-like 1; EGF-TM7; Emr1; EMR1 hormone receptor; F4/80; Gpf480; Ly71; lymphocyte antigen 71; TM7LN3 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| BM8 | |
| Flow Cytometry | |
| Brilliant Ultraviolet 805 | |
| Rat | |
| Mouse | |
| Q61549 | |
| 13733 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| ADGRE1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title