missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Factor X Monoclonal Antibody (156106)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA560900
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
F10 undergoes multiple processing steps before its preproprotein is converted to a mature two-chain form by the excision of the tripeptide RKR. Two chains of the factor are held together by 1 or more disulfide bonds; the light chain contains 2 EGF-like domains, while the heavy chain contains the catalytic domain which is structurally homologous to those of the other hemostatic serine proteases. The mature factor is activated by the cleavage of the activation peptide by factor IXa (in the intrisic pathway), or by factor VIIa (in the extrinsic pathway). The activated factor then converts prothrombin to thrombin in the presence of factor Va, Ca+2, and phospholipid during blood clotting. Mutations of this gene result in factor X deficiency, a hemorrhagic condition of variable severity.
Specifications
| Factor X | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P00742 | |
| F10 | |
| Mouse myeloma cell line NS0-derived recombinant human Coagulation Factor X, Leu24-Lys488 (predicted). | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot | |
| 156106 | |
| Unconjugated | |
| F10 | |
| Activated factor Xa heavy chain; blood coagulation factor X; Cf10; coagulation factor 10; coagulation factor X; coagulation factor X protein; F10; Factor 10; Factor X heavy chain; Factor X light chain; factor Xa; Factor10; FX; FXA; prothrombinase; RP11-98F14.9; stuart factor; Stuart-Prower factor; Unknown (protein for IMAGE:8121240); unnamed protein product | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 2159 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction