Få mere at vide
Invitrogen™ Factor X/Xa Monoclonal Antibody (5051)

Beskrivelse
This antibody detects the heavy chain of bovine Factor X and Xa, BEGRck factor Xa. It will cross-react with human, rabbit, sheep, porcine and canine factor X. This antibody does not recognize murine Factor X via Western blot. Inhibitory properties: inhibitory in clotting assay (APTT and PT), partial calcium dependence. Dissociation Constant: Kd = 9X10^(-11)M. Purity >95% by SDS-PAGE.
Tekniske data
Tekniske data
| Antigen | Factor X/Xa |
| Applications | Affinity Purification, Inhibition Assays, Radioimmune Assays (RIA), Western Blot |
| Classification | Monoclonal |
| Clone | 5051 |
| Concentration | 8.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50% water with 50% glycerol and no preservative |
| Gene | F10 |
| Gene Accession No. | O19045, P00742, P00743 |
| Gene Alias | Activated factor Xa heavy chain; blood coagulation factor X; Cf10; coagulation factor 10; coagulation factor X; coagulation factor X protein; F10; Factor X heavy chain; Factor X light chain; factor Xa; FX; FXA; prothrombinase; stuart factor; Stuart-Prower factor; Unknown (protein for IMAGE:8121240); unnamed protein product |
| Vis mere |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.