Learn More
Invitrogen™ Factor X/Xa Monoclonal Antibody (9050)

Description
This antibody detects mouse factor X (heavy chain) and mouse factor Xa (heavy chain) by ELISA and Western blot, and cross-reacts with human factor X/Xa (Western blot only). Western blot can be performed under reducing and non-reducing conditions. Inhibitory properties: Does not inhibit PT (mouse plasma). Purity >95% by SDS-PAGE.
Specifications
Specifications
| Antigen | Factor X/Xa |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 9050 |
| Concentration | 8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50% water with 50% glycerol and no preservative |
| Gene | F10 |
| Gene Accession No. | O88947, P00742 |
| Gene Alias | Activated factor Xa heavy chain; blood coagulation factor X; Cf10; coagulation factor 10; coagulation factor X; coagulation factor X protein; F10; Factor X heavy chain; Factor X light chain; factor Xa; FX; FXA; prothrombinase; stuart factor; Stuart-Prower factor; Unknown (protein for IMAGE:8121240); unnamed protein product |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.