missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Factor X/Xa (Calcium-dependent) Monoclonal Antibody (CaFX-50), Biotin
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA517644

Product Code. 15313931

  • 475.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is specific for Human Platelet Factors X and Xa in the presence of 5-10 mM CaCl2.

Factor X undergoes multiple processing steps before its preproprotein is converted to a mature two-chain form by the excision of the tripeptide RKR. Two chains of the factor are held together by 1 or more disulfide bonds; the light chain contains 2 EGF-like domains, while the heavy chain contains the catalytic domain which is structurally homologous to those of the other hemostatic serine proteases. The mature factor is activated by the cleavage of the activation peptide by factor IXa (in the intrisic pathway), or by factor VIIa (in the extrinsic pathway). The activated factor then converts prothrombin to thrombin in the presence of factor Va, Ca+2, and phospholipid during blood clotting. Mutations of this gene result in factor X deficiency, a hemorrhagic condition of variable severity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Factor X/Xa (Calcium-dependent)
Monoclonal
0.1 mg/mL
PBS with 4-5mg/mL BSA and 0.02% sodium azide
P00742
F10
Human Factor X purified from the supernatant of thrombin activated platelets.
100 μg
Primary
Human
Antibody
IgG1
ELISA, Western Blot
CaFX-50
Biotin
F10
Activated factor Xa heavy chain; blood coagulation factor X; Cf10; coagulation factor 10; coagulation factor X; coagulation factor X protein; F10; Factor X heavy chain; Factor X light chain; factor Xa; FX; FXA; prothrombinase; stuart factor; Stuart-Prower factor; Unknown (protein for IMAGE:8121240); unnamed protein product
Mouse
Protein G
RUO
2159
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.